missing translation for 'onlineSavingsMsg'
Learn More

D-Penicillamine, TRC

Product Code. 30011971
Change view
Click to view available options
Quantity:
25 mg
1 g
50 g
Unit Size:
1g
25mg
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011971 25 mg 25mg
30009893 1 g 1g
30003722 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011971 Supplier Toronto Research Chemicals Supplier No. TRCP22300025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-67-5
Molecular Formula C5 H11 N O2 S
Molecular Weight (g/mol) 149.21
Synonym Penicillamine, (2S)-2-Amino-3-methyl-3-sulfanylbutanoic acid, D-Valine, 3-mercapto-, Valine, 3-mercapto-, D- (8CI), (-)-Penicillamine, (2S)-2-Amino-3-methyl-3-sulfanylbutanoic acid, (S)-3,3-Dimethylcysteine, (S)-Penicillamine, 2-Amino-3-mercapto-3-methylbutanoic acid, Artamin, Cuprenil, Cuprimine, Cupripen, D-(-)-Penicillamine, D-3-Mercaptovaline, D-Mercaptovaline, D-Penamine, D-Penicillamine, DPA, Depamine, Depen, Kuprenil, Mercaptyl, Metalcaptase, NSC 81549, Pendramine, Penicillamin, Penicillamine, Perdolat, Reduced D-penicillamine, Reduced penicillamine, Sufirtan, Trolovol, β-Thiovaline
IUPAC Name (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
SMILES CC(C)(S)[C@@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C5H11NO2S/c1-5(2,9)3(6)4(7)8/h3,9H,6H2,1-2H3,(H,7,8)/t3-/m0/s1
Quantity 25 mg
Formula Weight 149.051
Shipping Condition Room Temperature
Chemical Name or Material Penicillamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.