missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Penicillamine, 99%, specified according to Ph.Eur. and USP

Product Code. p-178760 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10061450

Brand: Thermo Scientific Chemicals 129740050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52-67-5 | C5H11NO2S | 149.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-67-5
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00064302
InChI Key VVNCNSJFMMFHPL-VKHMYHEASA-N
Synonym d-penicillamine, penicillamine, cuprimine, d---penicillamine, depen, 3-mercapto-d-valine, cuprenil, d-penamine, --penicillamine, d-mercaptovaline
PubChem CID 5852
ChEBI CHEBI:7959
IUPAC Name (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid
SMILES CC(C)(C(C(=O)O)N)S

Specifications

Absorbance 0.07 max. (c=0.2, H2O, 1 cm cell) at 268nm
Melting Point 212.0°C
Color White
pH 4.5 to 5.5 (1% soln.)
Ignition Residue 0.1% max
Loss on Drying 0.5% max. (1 g, 60°C, 670 Pa) (P2O5)
Infrared Spectrum Authentic
Assay Percent Range 97.0 to 102% (USP)
Packaging Glass bottle
Linear Formula (CH3)2C(SH)CH(NH2)CO2H
Quantity 5 g
Merck Index 15, 7197
Specific Rotation Condition −65° (20°C c=5,1M NaOH,on dry)
Specific Rotation − 65.00
Solubility Information Solubility in water: 11.1% (20°C). Other solubilities: slightly soluble in ethanol,insoluble in ether,benzene,acetone,carbon tetra,chloride
Formula Weight 149.21
Percent Purity 99%
Physical Form Powder
Chemical Name or Material D(-)-Penicillamine, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • D(-)-Penicillamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
  • Reproductive toxicity Category 1B
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.