missing translation for 'onlineSavingsMsg'
Learn More

D-Pantothenic acid hemicalcium salt, MedChemExpress

Product Code. 16471478
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471478 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471478 Supplier Medchem Express Supplier No. HYN0681
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress D-Pantothenic acid hemicalcium salt (Vitamin B5 calcium salt), a vitamin, can reduce the patulin content of the apple juice.

D-Pantothenic acid hemicalcium salt (Vitamin B5 calcium salt), a vitamin, can reduce the patulin content of the apple juice.

Specifications

Chemical Name or Material D-Pantothenic acid hemicalcium salt
CAS 137-08-6
Color White
Purity Grade Notes Research
Molecular Formula C9H16NO5.1/2Ca
Quantity 5 g
Synonym Calcium pantothenate Calcium D-pantothenate Vitamin B5 calcium salt
Solubility Information H2O : 100 mg/mL (419.69 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (419.69 mM)
SMILES O[C@@H](C(NCCC([O-])=O)=O)C(C)(C)CO.[0.5Ca2+]
Molecular Weight (g/mol) 238.27
Formula Weight 238.27
Percent Purity 95.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.