missing translation for 'onlineSavingsMsg'
Learn More

D-Pantothenic Acid, Calcium Salt, MP Biomedicals™

Product Code. 11403190 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11403190 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11403190 Supplier MP Biomedicals Supplier No. 0210122890
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

D-Pantothenic acid is an essential vitamin (except in horses, ruminants).

Specifications

CAS 137-08-6
Physical Form Crystalline Powder
Quantity 500 g
Molecular Formula C18H32CaN2O10
MDL Number MFCD00002766
Synonym calcium pantothenate, d-pantothenic acid hemicalcium salt, d-pantothenic acid, calcium salt, pantothenic acid, calcium salt, d, r-n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-beta-alanine calcium salt, beta-alanine, n-2,4-dihydroxy-3,3-dimethyl-1-oxobutyl-, calcium salt, r
Solubility Information Soluble in water (1g/2.8mL; pH of a 5% aqueous solution is 7.2-8.0; CO2 free water: 8.7), glycerol; slightly soluble in ethanol or acetone.
InChI Key FAPWYRCQGJNNSJ-DXHDTSSINA-L
SMILES [Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O
IUPAC Name calcium bis(3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanoate)
Molecular Weight (g/mol) 476.54
PubChem CID 131847364
Formula Weight 238.3
Grade USP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.