missing translation for 'onlineSavingsMsg'
Learn More

D-Pantothenic acid calcium salt, 98%

Product Code. 11417957
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11417957

Brand: Thermo Scientific Alfa Aesar A16609.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 137-08-6 | C18H34CaN2O11 | 494.551 g/mol

Vitamin B-5D-Pantothenic acid calcium salt hydrate acts as a precursor in the biosynthesis of coenzyme A. It also involved in the catabolism of fatty acids, preparation of heme and the neurotransmitter acetylcholine. It is involved in the regulation of gene expression and in signal transduction. Further, it is used in sphingolipids, cholesterol and steroid hormones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vitamin B-5D-Pantothenic acid calcium salt hydrate acts as a precursor in the biosynthesis of coenzyme A. It also involved in the catabolism of fatty acids, preparation of heme and the neurotransmitter acetylcholine. It is involved in the regulation of gene expression and in signal transduction. Further, it is used in sphingolipids, cholesterol and steroid hormones.

Solubility
Soluble in water.

Notes
Hygroscopic. Air, light and moisture sensitive. Incompatible with strong oxidizing agents. Store in a cool place. Keep the container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-08-6
Molecular Formula C18H34CaN2O11
Molecular Weight (g/mol) 494.551
MDL Number MFCD00002766
InChI Key KUKRZUPDYACRAM-GXIDORRSSA-L
Synonym calcium 3-2,4-dihydroxy-3,3-dimethylbutanamido propanoate hydrate, d-pantothenic acid calcium salt hydrate, calcium hydrate dipantothenate, calcium bis 3-2,4-dihydroxy-3,3-dimethylbutanoyl amino propanoate hydrate
PubChem CID 16211719
IUPAC Name calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoate;hydrate
SMILES CC(C)(CO)C(C(=O)NCCC(=O)[O-])O.CC(C)(CO)C(C(=O)NCCC(=O)[O-])O.O.[Ca+2]

Specifications

Melting Point ∼138°C (decomposition)
Beilstein 3769272
Merck Index 14,7015
Solubility Information Soluble in water.
Quantity 500 g
Formula Weight 476.54
Optical Rotation +27° (c=5 in Water)
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material D-Pantothenic acid calcium salt hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.