missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Pantolactone, 99%

Product Code. 11322827
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322827 25 g 25g
11312827 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322827 Supplier Thermo Scientific Alfa Aesar Supplier No. L15983.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 599-04-2 | C6H10O3 | 130.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-04-2
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00005392
InChI Key SERHXTVXHNVDKA-BYPYZUCNSA-N
Synonym d---pantolactone, r-pantolactone, d-pantolactone, r-pantoyl lactone, d---pantoyl lactone, pantothenic lactone, r-3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, unii-j288d7o0js, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl-, 3r, r---pantolactone
PubChem CID 439368
ChEBI CHEBI:16719
IUPAC Name (3R)-3-hydroxy-4,4-dimethyloxolan-2-one
SMILES CC1(COC(=O)C1O)C

Specifications

Melting Point 89°C to 91°C
Boiling Point 120°C to 122°C (15 mmHg)
Quantity 25 g
Beilstein 80957
Sensitivity Hygroscopic
Merck Index 14,7013
Optical Rotation −51° (c=3 in Water)
Formula Weight 130.14
Percent Purity 99%
Chemical Name or Material D-(-)-Pantolactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.