missing translation for 'onlineSavingsMsg'
Learn More

D-Panthenol, TRC

Product Code. 30427849
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427849 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427849 Supplier Toronto Research Chemicals Supplier No. TRCP18030050G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Panthenol
CAS 81-13-0
Purity Grade Notes HPLC
Molecular Formula C9 H19 N O4
Quantity 50 g
Synonym (2R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide, (R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide, D-(+)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide, (+)-Panthenol, Alcopan 250, Bepanthen, Bepanthene, Bepantol, Cornergel, Cozyme, D(+)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide, D(+)-Panthenol, D(+)-Pantothenyl alcohol, D(+)-alpha,gamma-Dihydroxy-N-(3-hydroxypropyl)-beta,beta-dimethylbutyramide, D-P-A Injection, D-Panthenol, D-Pantothenyl alcohol, Dexpanthenol, Dolobene, Ilopan, Intrapan, Motilyn, NSC 302962, Panadon, Pantenyl, Panthenol, Panthenol 50W, (+)-Panthenol, Panthoderm, Pantol, Pantothenol, Pantothenyl alcohol, N-Pantoylpropanolamine, Provitamin B, Provitamin B5, Synapan, Thenalton, Urupan, Zentinic, d-Panthenol, d-Panthenol 50, d-Pantothenol, d-Pantothenyl alcohol
InChI Formula InChI=1S/C9H19NO4/c1-9(2,6-12)7(13)8(14)10-4-3-5-11/h7,11-13H,3-6H2,1-2H3,(H,10,14)/t7-/m0/s1
SMILES CC(C)(CO)[C@@H](O)C(=O)NCCCO
IUPAC Name (2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
Molecular Weight (g/mol) 205.25
Formula Weight 205.1314
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.