missing translation for 'onlineSavingsMsg'
Learn More

D-Ornithine Hydrochloride, TRC

Product Code. 17919734
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17919734 1 g 1g
17968214 5 g 5g
17907914 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17919734 Supplier Toronto Research Chemicals Supplier No. TRCO6955451G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16682-12-5
Molecular Formula C5 H12 N2 O2 . Cl H
Molecular Weight (g/mol) 168.6219
Synonym D-Ornithine, hydrochloride (1:1), D-Ornithine, monohydrochloride (9CI), Ornithine, monohydrochloride, D- (8CI), (R)-Ornithine hydrochloride, D-(-)-2,5-diaminopentanoic acid hydrochloride, (R)-2,5-Diaminopentanoic Acid Hydrochloride, D-Ornithine Monohydrochloride, D-(-)-Ornithine Monohydrochloride, (D)-α,δ-Diaminovaleric Acid, 5-Amino-D-norvaline
IUPAC Name (2R)-2,5-diaminopentanoic acid;hydrochloride
SMILES Cl.NCCC[C@@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m1./s1
Quantity 1 g
Formula Weight 168.0666
Shipping Condition Room Temperature
Chemical Name or Material D-Ornithine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.