missing translation for 'onlineSavingsMsg'
Learn More

D-Ornithine hydrochloride, 98+%

Product Code. 11310224
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11310224 5 g 5g
11300224 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11310224 Supplier Thermo Scientific Alfa Aesar Supplier No. L00793.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16682-12-5 | C5H13ClN2O2 | 168.621 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16682-12-5
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.621
MDL Number MFCD00012917
InChI Key GGTYBZJRPHEQDG-PGMHMLKASA-N
Synonym d-ornithine monohydrochloride, d-ornithine hydrochloride, d-ornithine hcl, h-d-orn-oh.hcl, r-ornithine hydrochloride, d-ornithine, monohydrochloride, l---ornithine hydrochloride, r-2,5-diaminopentanoic acid hydrochloride, 2r-2,5-diaminopentanoic acid hydrochloride, ornithine hcl
PubChem CID 443122
ChEBI CHEBI:4221
IUPAC Name (2R)-2,5-diaminopentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN.Cl

Specifications

Melting Point 237°C (decomposition)
Beilstein 4153339
Quantity 5 g
Formula Weight 168.62
Optical Rotation −23° (c=5 in 5N HCl)
Percent Purity ≥98%
Chemical Name or Material D-Ornithine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.