missing translation for 'onlineSavingsMsg'
Learn More

D-NG-Nitroarginine Methyl Ester Hydrochloride, TRC

Product Code. 30384680
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384680 500 mg
30376387 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384680 Supplier Toronto Research Chemicals Supplier No. TRCD463578500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50912-92-0
Molecular Formula C7H15N5O4 . (HCl)
Molecular Weight (g/mol) 233.23
Synonym N5-[Imino(nitroamino)methyl]-D-ornithine Methyl Ester Hydrochloride, N5-[Imino(nitroamino)methyl]-D-ornithine Methyl Ester Monohydrochloride
IUPAC Name methyl (2R)-2-amino-5-[(N-nitrocarbamimidoyl)amino]pentanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](N)CCCNC(=N)NN(=O)=O

Specifications

InChI Formula InChI=1S/C7H15N5O4.ClH/c1-16-6(13)5(8)3-2-4-10-7(9)11-12(14)15;/h5H,2-4,8H2,1H3,(H3,9,10,11);1H/t5-;/m1./s1
Quantity 500 mg
Formula Weight 269.0891
Shipping Condition Room Temperature
Chemical Name or Material D-NG-Nitroarginine Methyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.