missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(D)-(+)-Neopterin, Tocris Bioscience™
Description
Precursor of biopterin synthesis. Synthesized in response to interferon-γ stimulation; used as a marker of T helper cell-induced immune activation. Also an indicator of oxidative stress; modulates the effects of reactive oxygen species (ROS).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 253.21 |
| Percent Purity | >98% |
| Chemical Name or Material | (D)-(+)-Neopterin |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?