missing translation for 'onlineSavingsMsg'
Learn More

D-Myo-inositol, 99.26%, For HPLC analysis, MP Biomedicals™

Product Code. 11471881 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471881 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11471881 Supplier MP Biomedicals Supplier No. 0219468890
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 87-89-8
Color White
Density 6.2g/mL
Assay Percent Range 99.26%
Molecular Formula C6H12O6
Quantity 500 g
Synonym scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol
InChI Key CDAISMWEOUEBRE-UHFFFAOYSA-N
SMILES C1(C(C(C(C(C1O)O)O)O)O)O
IUPAC Name cyclohexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol) 180.156
PubChem CID 892
ChEBI CHEBI:24848
Formula Weight 180.2
Physical Form Powder
Chemical Name or Material D-Myo-inositol, For HPLC analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.