missing translation for 'onlineSavingsMsg'
Learn More

D-myo-Inositol-3-phosphate sodium salt, 98%, Thermo Scientific Chemicals

Product Code. 15468619
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
1 mg
Unit Size:
0.1mg
0.5mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468619 0.1 mg 0.1mg
15458619 0.5 mg 0.5mg
15478619 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15468619 Supplier Thermo Scientific Alfa Aesar Supplier No. J67072.!Z
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD05864254
Synonym 3-IP1 sodium salt; Ins(3)P1 sodium salt

Specifications

Color White
Quantity 0.1 mg
Solubility Information Soluble in water
Formula Weight 282.12
Percent Purity 98%
Physical Form Lyophilized Powder
Chemical Name or Material D-myo-Inositol-3-phosphate sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.