missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Melibiose monohydrate, 99+%

Product Code. p-4509787 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10745961 25 g Glass bottle 25g
10307010 100 g Glass bottle 100g
11976891 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10745961 Supplier Thermo Scientific Chemicals Supplier No. 125370250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 66009-10-7 | C12H24O12 | 360.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66009-10-7
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
MDL Number MFCD00198188
InChI Key CHIDEFLSUMQFBY-UHFFFAOYNA-N
Synonym d +-melibiose monohydrate, d-+-melibiose hydrate, 585-99-9 anhydrous, melibiose hplc, 6-, a-d-galactopyranosyl-d-glucopyranose, 6-o-a-d-galactopyranosyl-d-glucopyranose hydrate, 6-o-a-d-galactopyranosyl-d-glucose monohydrate, 6-o-a-d-galactopyranosyl-d-glucopyranose monohydrate, 6-o-alpha-d-galactopyranosyl-d-glucose monohydrate, d-glucose, 6-o-a-d-galactopyranosyl-, monohydrate 9ci
PubChem CID 71308738
IUPAC Name (2R,3S,4R,5R)-2,3,4,5-tetrahydroxy-6-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanal;hydrate
SMILES O.OCC1OC(OCC(O)C(O)C(O)C(O)C=O)C(O)C(O)C1O

Specifications

Acidity 0.4mL 0.1N NaOH/6g max.
Melting Point 182.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Odor Odorless
Packaging Glass bottle
Quantity 25 g
Beilstein 31,421
Merck Index 14,582
Specific Rotation Condition + 135.00 (20.00°C c=4,H2O/NH3)
Specific Rotation + 135.00
Solubility Information Solubility in water: soluble
Formula Weight 360.32
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Melibiose monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.