missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Mannose, MP Biomedicals

Product Code. 11493470 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493470 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11493470 Supplier MP Biomedicals Supplier No. 0210225090
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in the formation of glycan structure and glycosylation, and can be utilized by the brain as an alternative energy source.

  • D-(+)-Mannose, a C-2 epimer of D-glucose, is used in the formation of glycan structure and glycosylation
  • D-Mannose has been used in a study to assess the synthesis of a family of amphiphilic glycopolymers
  • It has also been used in a study to investigate the early detection of bronchiolitis obliterans after lung transplantation

Specifications

CAS 3458-28-4
Melting Point 129°C to 140°C
Color White
Boiling Point 527.1°C (760 mmHg)
Molecular Formula C6H12O6
Linear Formula C6H12O6
MDL Number MFCD00064122
Quantity 500 g
Synonym 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers
Solubility Information Soluble in water (50 gm/mL-clear colorless solution).
Molecular Weight (g/mol) 180.156
Formula Weight 180.2
Percent Purity ≥98%
Grade Not Available
Physical Form Powder
Chemical Name or Material D-(+)-MANNOSE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.