missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Mannose, Extra Pure, SLR, Fisher Chemical

Product Code. 10386090 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10386090 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10386090 Supplier Fisher Chemical Supplier No. M/2420/45
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

10g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

CAS 3458-28-4
Melting Point 133°C
Color White
Packaging Plastic powder jar
Molecular Formula C6H12O6
MDL Number 64122
Quantity 10 g
Formula Weight 180.16g/mol
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.