missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Mannose, 99+%

Product Code. 10726371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
1kg
2.5kg
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10726371 1 kg Plastic bottle 1kg
10466790 25 g Plastic bottle 25g
10265340 100 g Plastic bottle 100g
11926901 2.5 kg Plastic drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10726371 Supplier Thermo Scientific Chemicals Supplier No. 150600010
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3458-28-4 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3458-28-4
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
Synonym 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers

Specifications

Melting Point 133.0°C
Color White
Loss on Drying 0.45% max.
Infrared Spectrum Authentic
Assay Percent Range 98% min. (ELSD) (HPLC)
Odor Odorless
Packaging Plastic bottle
Quantity 1 kg
Beilstein 31284
Merck Index 155812
Specific Rotation Condition + 14.00 (20.00°C 589nm,c=10,H2O)
Specific Rotation + 14.00
Solubility Information Solubility in water: 2480g/L (17°C)
Formula Weight 180.16
Percent Purity 99+%
Physical Form Crystals or Powder
Chemical Name or Material D(+)-Mannose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.