missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Mannose, 99+%

Product Code. 10265340 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
1kg
2.5kg
25g
This item is not returnable. View return policy

Product Code. 10265340

Brand: Thermo Scientific Chemicals 150601000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 3458-28-4 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3458-28-4
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
Synonym 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers

Specifications

Melting Point 133.0°C
Color White
Loss on Drying 0.45% max.
Infrared Spectrum Authentic
Assay Percent Range 98% min. (ELSD) (HPLC)
Odor Odorless
Packaging Plastic bottle
Quantity 100 g
Beilstein 31284
Merck Index 155812
Specific Rotation Condition + 14.00 (20.00°C 589nm,c=10,H2O)
Specific Rotation + 14.00
Solubility Information Solubility in water: 2480g/L (17°C)
Formula Weight 180.16
Percent Purity 99+%
Physical Form Crystals or Powder
Chemical Name or Material D(+)-Mannose
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.