missing translation for 'onlineSavingsMsg'
Learn More

D-Mannitol, TRC

Product Code. 30471563
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471563 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471563 Supplier Toronto Research Chemicals Supplier No. TRCM16500025G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Mannitol
CAS 69-65-8
Molecular Formula C6 H14 O6
Quantity 25 g
Synonym Mannitol, d-Mannitol, D-Mannitol, Cordycepic acid (6CI,7CI), Mannitol, D- (8CI), Brightmoon, Bronchitol, Cerestar 16700, D-(-)-Mannitol, D-Mannit, Diosmol, E 421, Isotol, Manicol, Maniton S, Manna sugar, Mannidex, Mannidex 16700, Mannigen, Mannistol, Mannit, Mannit 60, Mannit P, Mannit S, Mannite, Mannitol 35, Mannitol 60, Mannitol P 60, Mannitol SD 200, Mannitolum, Mannogem 2080, Mannogem EZ, Marine Crystal, Nonpareil 108, Nonpareil 108(200), Orocell 200, Osmitrol, Osmosal, Parteck 300, Parteck Delta M, Parteck M 100, Parteck M 200, Parteck M 300, Partek M, Pearlitol, Pearlitol 100, Pearlitol 100SD, Pearlitol 160C, Pearlitol 2005D, Pearlitol 200SD, Pearlitol 25C, Pearlitol 50C, Pearlitol SD 100, Perlitol SD 200, Perteck M, PharmMannidex, PharmMannidex 16701, Resectisol
InChI Formula InChI=1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
IUPAC Name (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol) 182.17
Formula Weight 182.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.