missing translation for 'onlineSavingsMsg'
Learn More

D-Mannitol, MedChemExpress

Product Code. 16401398
Encompass_Preferred
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 16401398

Brand: Medchem Express HYN0378

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D-Mannitol is an osmotic diuretic agent and a weak renal vasodilator.

D-Mannitol is an osmotic diuretic agent and a weak renal vasodilator.

Specifications

Chemical Name or Material D-Mannitol
CAS 69-65-8
Color White
Purity Grade Notes Research
Molecular Formula C6H14O6
Quantity 5 g
Synonym Mannitol Mannite
Solubility Information H2O : ≥ 36 mg/mL (197.62 mM)
SMILES OC[C@H]([C@H]([C@@H]([C@@H](CO)O)O)O)O
Molecular Weight (g/mol) 182.17
Formula Weight 182.17
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.