missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Mannitol, MedChemExpress
Description
D-Mannitol is an osmotic diuretic agent and a weak renal vasodilator.
Specifications
Specifications
| Chemical Name or Material | D-Mannitol |
| CAS | 69-65-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C6H14O6 |
| Quantity | 10 g |
| Synonym | Mannitol Mannite |
| Solubility Information | H2O : ≥ 36 mg/mL (197.62 mM) |
| SMILES | OC[C@H]([C@H]([C@@H]([C@@H](CO)O)O)O)O |
| Molecular Weight (g/mol) | 182.17 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?