missing translation for 'onlineSavingsMsg'
Learn More

D-Mannitol, For ACS analysis, 98.4%, MP Biomedicals™

Product Code. 11452872 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452872 1 kg 1kg
11432872 250 g 250g
11442872 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452872 Supplier MP Biomedicals Supplier No. 0215254091
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Color White
Density 1.52g/mL
Assay Percent Range 0.984
Quantity 1 kg
Formula Weight 182.2
Physical Form Powder
Chemical Name or Material D-Mannitol, For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.