missing translation for 'onlineSavingsMsg'
Learn More

D-Mannit 1000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30311870
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30311870 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30311870

Brand: Dr. Ehrenstorfer DREA14752000ME1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Mannitol
CAS 69-65-8
Analyte or Component Names D-Mannitol
Shipping Condition Room Temperature
Molecular Formula C6 H14 O6
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Mannitol, d-Mannitol, D-Mannitol, Cordycepic acid (6CI,7CI), Mannitol, D- (8CI), Brightmoon, Bronchitol, Cerestar 16700, D-(-)-Mannitol, D-Mannit, Diosmol, E 421, Isotol, Manicol, Maniton S, Manna sugar, Mannidex, Mannidex 16700, Mannigen, Mannistol, Mannit, Mannit 60, Mannit P, Mannit S, Mannite, Mannitol 35, Mannitol 60, Mannitol P 60, Mannitol SD 200, Mannitolum, Mannogem 2080, Mannogem EZ, Marine Crystal, Nonpareil 108, Nonpareil 108(200), Orocell 200, Osmitrol, Osmosal, Parteck 300, Parteck Delta M, Parteck M 100, Parteck M 200, Parteck M 300, Partek M, Pearlitol, Pearlitol 100, Pearlitol 100SD, Pearlitol 160C, Pearlitol 2005D, Pearlitol 200SD, Pearlitol 25C, Pearlitol 50C, Pearlitol SD 100, Perlitol SD 200, Perteck M, PharmMannidex, PharmMannidex 16701, Resectisol, Lactitol Monohydrate Impurity C, Sorbitol Impurity A, myo-Inositol Impurity A, Lactitol Impurity C
InChI Formula InChI=1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
IUPAC Name (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
Solvent or Matrix Methanol
Molecular Weight (g/mol) 182.17
Quantity 1 mL
Formula Weight 182.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.