Learn More
D-(+)-Malic acid, 98+%
Description
D-(+)-Malic acid used as an acidulant and flavoring agent, food additive. And it is also used in the place of the less sour citric acid in sour sweets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
D-(+)-Malic acid used as an acidulant and flavoring agent, food additive. And it is also used in the place of the less sour citric acid in sour sweets.
Solubility
Soluble in methanol, ethanol, acetone, ether.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
| Melting Point | 100°C to 104°C |
| Density | 1.6 |
| Quantity | 25 g |
| Beilstein | 1723540 |
| Merck Index | 14,5707 |
| Solubility Information | Soluble in methanol,ethanol,acetone,ether. |
| Optical Rotation | +28° (c=5.5 in pyridine) |
| Formula Weight | 134.09 |
| Percent Purity | ≥98% |
| Chemical Name or Material | D- (+)-Malic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.