missing translation for 'onlineSavingsMsg'
Learn More

D-Lysine, Thermo Scientific™

Product Code. 17730719 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17730719 5 g 5g
17720719 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17730719 Supplier Thermo Scientific Acros Supplier No. 469950050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material D-Lysine
Melting Point 218°C
CAS 923-27-3
Boiling Point 311°C
Color Beige to White
Infrared Spectrum Conforms
Molecular Formula C6 H14 N2 O2
Packaging Glass Bottle
MDL Number 00008234
Specific Rotation -12° to -16°
Synonym (R)-2,6-Diaminohexanoic acid
Water ≤5 %
Alpha Vector LYSINE
Specific Rotation Condition (20°C, 589 nm) (c=1 aq. soln.)
Molecular Weight (g/mol) 146.19
Quantity 5 g
Physical Form Crystalline powder
Product Line Life science
Shipping Condition No special requirements
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.