missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Lysine hydrochloride, 99+%

Product Code. p-3776608 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass Bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10125150 1 g Glass Bottle 1g
10308520 10 g Glass Bottle 10g
10435295 50 g Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10125150 Supplier Thermo Scientific Chemicals Supplier No. 243080010
Add to basket
This item is not returnable. View return policy

CAS: 7274-88-6 | C6H15ClN2O2 | 182.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7274-88-6
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00012920
InChI Key BVHLGVCQOALMSV-XZNNNFJINA-N
Synonym d-lysine hydrochloride, d-lysine monohydrochloride, d-lysine hcl, d-lysine, monohydrochloride, h-d-lys-oh.hcl, d-lysine, hydrochloride, unii-kk611jde1n, lysine, monohydrochloride, d, r-2,6-diaminohexanoic acid hydrochloride, lysine, hydrochloride, d
PubChem CID 81691
ChEBI CHEBI:53558
SMILES Cl.NCCCC[C@@H](N)C(O)=O

Specifications

Arsenic (As) 1ppm max.
Color White
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Assay Percent Range 99+%
Loss on Drying 0.3% max. (105°C, 3 hrs)
Packaging Glass Bottle
Linear Formula H2N(CH2)4CH(NH2)COOH·HCl
Specific Rotation -20.5° to -21.5° (20°C, 589nm) (c=8, 6N HCl)
Fieser 01,959; 07,79; 01,977
Sulfated Ash 0.1% max.
IUPAC Name (2R)-2,6-diaminohexanoic acid hydrochloride
Specific Rotation Condition −21° (20°C c=8,6N HCl)
Quantity 1 g
Formula Weight 182.65
Percent Purity 99.0 to 101.0%
Physical Form Crystalline Powder
Chemical Name or Material D-Lysine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.