missing translation for 'onlineSavingsMsg'
Learn More

D-Lysine(>90%), TRC

Product Code. 30135640
Change view
Click to view available options
Quantity:
2.5 g
10 g
25 g
Unit Size:
10g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135640 25 g 25g
30133908 10 g 10g
30003886 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135640 Supplier Toronto Research Chemicals Supplier No. TRCL46893025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 923-27-3
Molecular Formula C6H14N2O2
Molecular Weight (g/mol) 146.19
Synonym (R)-Lysine;
IUPAC Name 2,4-dimethylphenol
SMILES [C@@H](CCCCN)(C(O)=O)N

Specifications

InChI Formula InChI=1S/C8H10O/c1-6-3-4-8(9)7(2)5-6/h3-5,9H,1-2H3
Quantity 25 g
Formula Weight 146.11
Shipping Condition Room Temperature
Chemical Name or Material D-Lysine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.