missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress D-Lys(Z)-Pro-Arg-pNA diacetate

Product Code. 16454967
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16454967 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16454967 Supplier MedChemExpress Supplier No. HYP0021A
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress D-Lys(Z)-Pro-Arg-pNA diacetate (Spectrozyme PCa) is a chromogenic substrate.

Specifications

CAS 108963-70-8
Solubility H2O : 2 mg/mL (2.58 mM∣Need ultrasonic)
Synonym Spectrozyme PCa∣ Chromozym Pca diacetate
Content And Storage Sealed storage, away from moisture and light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
For Use With (Application) Neuroscience-Neuromodulation
Quantity 50 mg
Molecular Weight (g/mol) 773.83 g/mol
Product Type Dye Reagents
Purity or Quality Grade 95.82%
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.