missing translation for 'onlineSavingsMsg'
Learn More

D-Luciferin sodium salt monohydrate, 98+%

Product Code. 10758942 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass bottle
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10758942 50 mg Glass bottle 50mg
10668254 10 mg Glass bottle 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10758942 Supplier Thermo Scientific Acros Supplier No. 336780500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103404-75-7 | C11H7N2NaO3S2 | 302.298 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103404-75-7
Molecular Formula C11H7N2NaO3S2
Molecular Weight (g/mol) 302.298
MDL Number MFCD11865368
InChI Key BZNVUYVALNTPBG-QWLWRJJTSA-M
Synonym Firefly Luciferin monosodium salt monohydrate
PubChem CID 133109097
IUPAC Name sodium;(2E,4R)-2-(6-oxo-1,3-benzothiazol-2-ylidene)-1,3-thiazolidine-4-carboxylate
SMILES C1C(NC(=C2N=C3C=CC(=O)C=C3S2)S1)C(=O)[O-].[Na+]

Specifications

Assay Percent Range 98+%
Infrared Spectrum Authentic
Formula Weight 320.32
Percent Purity ≥98.0%
Packaging Glass bottle
Specific Extinction 17500 min. (385nm, 0.5M Na2CO3) min. (pH 11.5)
Color Yellow
Quantity 50 mg
Physical Form Powder
Chemical Name or Material D-Luciferin sodium salt monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.