missing translation for 'onlineSavingsMsg'
Learn More

Melford D-Luciferin Sodium Salt

Product Code. 30018446
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018446 1 g 1g
30018444 100 mg 100mg
30018443 50 mg 50mg
30018445 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30018446 Supplier Melford Supplier No. L370801.0
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

RPI offers the highest purity D-Luciferin Sodium Salt for a wide range of bioluminescent applications.

RPI offers the highest purity D-Luciferin Sodium Salt for a wide range of bioluminescent applications. Allows highly sensitive bioluminescent measurements. Ideal for reporter gene assays, ATP assays, whole animal studied, DNA sequencing and more. Our product is specially purified and tested extensively to ensure the highest signal and reproducibility with low background and sensitivity. Guaranteed purity is determined by both standard HPLC and Chiral HPLC methods. Protect from light and moisture.

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.