missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ D-Luciferin sodium salt

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
D-Luciferin sodium salt is a firefly luciferase substrate (Km = ∽2 μM). Used in many standard bioluminescence imaging (BLI) techniques, including monitoring tumour growth, and investigating intracellular signaling activity in vitro and in vivo. Cell permeable. Emission maximum = 562 nm.
Specifications
Specifications
| Quantity | 50 mg |
| Molecular Formula | C11H7N2NaO3S2 |
| Product Type | D-Luciferin sodium salt |
| Purity | >99% |
| Molecular Weight (g/mol) | 302.3 |
| Inhibitors | D-Luciferin sodium salt |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction