missing translation for 'onlineSavingsMsg'
Learn More

BD D- Luciferin Potassium Salt

Product Code. 15888788
Click to view available options
Quantity:
10 mg
50 mg
100 mg
250 mg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15888788

Brand: BD 556888

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetically made substrate which can be catalyzed by firefly luciferase

  • Also known as beetle luciferin
  • Lyophilized in form of monopotassium salt
  • Purified by HPLC to remove dehydroluciferin and L-Luciferin
  • Form dehydroluciferin in presence of ultraviolet light and/or moisture

Luminometer

TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized
Type D-Luciferin
Quantity 250 mg
Content And Storage -20°C
For Use With (Application) Luminometer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.