Learn More
Melford D-Luciferin Potassium Salt
Product Code. 30018441
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018441 250 mg 250mg
30018442 1 g 1g
30018439 50 mg 50mg
30018440 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30018441 Supplier Melford Supplier No. L370600.25
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
RPI offers the highest purity D-Luciferin Potassium Salt for a wide range of bioluminescent applications.
RPI offers the highest purity D-Luciferin Potassium Salt for a wide range of bioluminescent applications. Produces bioluminescence when oxidized by firefly luciferase. Allows highly sensitive bioluminescent measurements. Common research uses reporter gene assays, ATP assays, whole animal studies, DNA sequencing and others. Our product is specially purified and tested extensively to ensure the highest signal and reproducibility with low background and sensitivity. Guaranteed purity is determined by both standard HPLC and Chiral HPLC methods. Protect from light and moisture.
Specifications
Quantity 250 mg