missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Luciferin, Thermo Scientific Chemicals

Catalog Number 10229463
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass bottle
2591-17-5
C11H8N2O3S2
280.3
MFCD00042929
IWJYWBVPCGUPLO-SSDOTTSWSA-N
luciferin, d-luciferin, s-2-6-hydroxybenzo d thiazol-2-yl-4,5-dihydrothiazole-4-carboxylic acid, luciferin firefly, s-2-6-hydroxy-2-benzothiazolyl-2-thiazoline-4-carboxylic acid, 4s-2-6-oxo-1,3-benzothiazol-2-ylidene-1,3-thiazolidine-4-carboxylic acid, 4-thiazolecarboxylicacid, 4,5-dihydro-2-6-hydroxy-2-benzothiazolyl-, 4s
7023066
(4S)-2-(6-oxo-1,3-benzothiazol-2-ylidene)-1,3-thiazolidine-4-carboxylic acid
C1C(NC(=C2N=C3C=CC(=O)C=C3S2)S1)C(=O)O
2591-17-5
C11H8N2O3S2
280.3
MFCD00042929
IWJYWBVPCGUPLO-SSDOTTSWSA-N
luciferin, d-luciferin, s-2-6-hydroxybenzo d thiazol-2-yl-4,5-dihydrothiazole-4-carboxylic acid, luciferin firefly, s-2-6-hydroxy-2-benzothiazolyl-2-thiazoline-4-carboxylic acid, 4s-2-6-oxo-1,3-benzothiazol-2-ylidene-1,3-thiazolidine-4-carboxylic acid, 4-thiazolecarboxylicacid, 4,5-dihydro-2-6-hydroxy-2-benzothiazolyl-, 4s
7023066
(4S)-2-(6-oxo-1,3-benzothiazol-2-ylidene)-1,3-thiazolidine-4-carboxylic acid
C1C(NC(=C2N=C3C=CC(=O)C=C3S2)S1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 200°C
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10 mg
Merck Index 15, 4117
Specific Rotation -32.0° to -36.0° (20°C, 589nm) (c=1, DMF)
Solubility Information Solubility in water: slighty soluble. Other solubilities: soluble in methanol
Formula Weight 280.3
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material D(-)-Luciferin
Show More Show Less
Product Identifier
  • D(-)-Luciferin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'