missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress D-Luciferin

Product Code. 16461467
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461467 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16461467 Supplier MedChemExpress Supplier No. HY12591A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D-Luciferin (D-(-)-Luciferin) is the substrate of luciferases that catalyze the production of light in bioluminescent insects.

Specifications

CAS 2591-17-5
Solubility DMSO : 50 mg/mL (178.37 mM∣Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
Synonym D-(-)-Luciferin∣ Firefly luciferin∣ Beetle Luciferin
Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Quantity 50 mg
Molecular Weight (g/mol) 280.32 g/mol
Product Type Dye Reagents
Purity or Quality Grade 98.14%
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.