missing translation for 'onlineSavingsMsg'
Learn More

D-Leucine, TRC

Product Code. 30470714
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30470714 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30470714 Supplier Toronto Research Chemicals Supplier No. TRCL3301502.5G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 328-38-1
Molecular Formula C6H13NO2
Synonym (R)-(-)-Leucine, (R)-4-Methyl-2-aminopentanoic Acid, (R)-Leucine, NSC 77687
InChI Formula InChI=1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m1/s1
SMILES CC(C)C[C@@H](N)C(=O)O
IUPAC Name (2R)-2-amino-4-methylpentanoic acid
Molecular Weight (g/mol) 131.17
Quantity 2.5 g
Formula Weight 131.0946
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material D-Leucine
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.