missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals α-D-Lactose monohydrate, ACS

Product Code. 11323268
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323268 2 kg 2kg
11313268 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323268 Supplier Thermo Scientific Chemicals Supplier No. 036218.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

μa-D-Lactose monohydrate, ACS, CAS # 64044-51-5, is the crystalline form of D-Lactose, which is the major sugar found in milk. | CAS: 5989-81-1 | C12H24O12 | 360.312 g/mol

It is also heavily used in the pharmaceutical industry as a pill filler. They are capable of fermenting lactose for ethanol production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• μa-D-Lactose monohydrate, ACS is the crystalline form of D-Lactose
• It is composed of galactose and glucose subunits

Application

• μa-D-Lactose monohydrate can be used to ferment lactose for ethanol production in laboratory settings
• It may be used as a reference standard for lactose quantitation

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5989-81-1
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.312
MDL Number MFCD00150747
InChI Key WSVLPVUVIUVCRA-ZEAJSSQNSA-N
Synonym d-lactose monohydrate, d-+-lactosemonohydrate
PubChem CID 133126686
IUPAC Name (3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O

Specifications

Melting Point ∼218°C (decomposition)
Physical Form Crystalline
Quantity 2 kg
Beilstein 3768231
Merck Index 14,5343
Solubility Information Soluble in water (warm) (0.1g/mL),and ethanol (very slightly soluble). Insoluble in chloroform,and ether.
Formula Weight 360.32 (342.30 Anhydrous)
Grade ACS
Density 1.525 g/mL
Chemical Name or Material alpha-D-Lactose monohydrate, For analysis ACS

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.