missing translation for 'onlineSavingsMsg'
Learn More

D-Lactic Acid, TRC

Product Code. 30483275
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483275 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483275 Supplier Toronto Research Chemicals Supplier No. TRCL113485250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Lactic Acid
CAS 10326-41-7
Molecular Formula C3H6O3
Quantity 250 mg
Synonym (-)-Lactic Acid, (2R)-2-Hydroxypropanoic Acid, (R)-(-)-Lactic Acid, (R)-2-Hydroxypropanoic Acid, (R)-2-Hydroxypropionic Acid, (R)-Lactic Acid, (R)-α-Hydroxypropionic Acid, D-(-)-Lactic Acid, D-Lactic Acid, l-Lactic Acid
InChI Formula InChI=1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m1/s1
SMILES O[C@@H](C)C(O)=O
IUPAC Name (2R)-2-hydroxypropanoic acid
Molecular Weight (g/mol) 90.08
Formula Weight 90.0317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.