missing translation for 'onlineSavingsMsg'
Learn More

D,L-threo-Droxidopa-13C2,15N Hydrochloride, TRC

Product Code. 30388009
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388009 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388009 Supplier Toronto Research Chemicals Supplier No. TRCD68995210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D,L-threo-Droxidopa-13C2,15N Hydrochloride
CAS 1329556-63-9
Molecular Formula C713C2H12Cl15NO5
Quantity 10 mg
Synonym (βS)-rel-β,3-Dihydroxy-D-tyrosine-13C2,15N Hydrochloride, DL-threo-3-(3,4-Dihydroxyphenyl)serine-13C2,15N Hydrochloride, DL-DOPS-13C2,15N, L-Threodops-13C2,15N Hydrochloride, DL-threo-3,4-Dihydroxyphenylserine- 13C2,15N Hydrochloride, DL-threo-DOPS-13C2,15N Hydrochloride, threo-2-Amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropionic Acid-13C2,15N Hydrochloride
InChI Formula InChI=1S/C9H11NO5.ClH/c10-7(9(14)15)8(13)4-1-2-5(11)6(12)3-4;/h1-3,7-8,11-13H,10H2,(H,14,15);1H/t7-,8+;/m0./s1/i7+1,9+1,10+1;
SMILES OC1=C(O)C=CC([C@@H](O)[13C@H]([15NH2])[13C](O)=O)=C1.Cl
Molecular Weight (g/mol) 252.63
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.