missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D,L-threo-Droxidopa-13C2,15N Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | D,L-threo-Droxidopa-13C2,15N Hydrochloride |
| CAS | 1329556-63-9 |
| Molecular Formula | C713C2H12Cl15NO5 |
| Quantity | 1 mg |
| Synonym | (βS)-rel-β,3-Dihydroxy-D-tyrosine-13C2,15N Hydrochloride, DL-threo-3-(3,4-Dihydroxyphenyl)serine-13C2,15N Hydrochloride, DL-DOPS-13C2,15N, L-Threodops-13C2,15N Hydrochloride, DL-threo-3,4-Dihydroxyphenylserine- 13C2,15N Hydrochloride, DL-threo-DOPS-13C2,15N Hydrochloride, threo-2-Amino-3-(3,4-dihydroxyphenyl)-3-hydroxypropionic Acid-13C2,15N Hydrochloride |
| InChI Formula | InChI=1S/C9H11NO5.ClH/c10-7(9(14)15)8(13)4-1-2-5(11)6(12)3-4;/h1-3,7-8,11-13H,10H2,(H,14,15);1H/t7-,8+;/m0./s1/i7+1,9+1,10+1; |
| SMILES | OC1=C(O)C=CC([C@@H](O)[13C@H]([15NH2])[13C](O)=O)=C1.Cl |
| Molecular Weight (g/mol) | 252.63 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?