missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D,L-Pantolactone, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | DL-Pantolactone |
| CAS | 79-50-5 |
| Molecular Formula | C6 H10 O3 |
| Quantity | 50 g |
| Synonym | D,L-Pantolactone, DL-Pantolactone, (±)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, 2(3H)-Furanone, dihydro-3-hydroxy-4,4-dimethyl-, 2(3H)-Furanone, dihydro-3-hydroxy-4,4-dimethyl-, (±)-, Butyric acid, α,γ-dihydroxy-β,β-dimethyl-, γ-lactone (4CI), Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, (RS)-Pantolactone, (±)-2-Hydroxy-3,3-dimethyl-γ-butyrolactone, (±)-Pantolactone, (±)-Pantoyl lactone, 2,4-Dihydroxy-3,3-dimethylbutanoic acid γ-lactone, 2,4-Dihydroxy-3,3-dimethylbutyric acid γ-lactone, 2-Hydroxy-3,3-dimethyl-γ-butyrolactone, 2-Hydroxy-3,3-dimethylbutan-4-olide, 3-Hydroxy-4,4-dimethyl-2-oxotetrahydrofuran, 3-Hydroxy-4,4-dimethyl-2-tetrahydrofuranone, 3-Hydroxy-4,4-dimethyl-dihydro-furan-2-one, 3-Hydroxy-4,4-dimethyldihydrofuran-2(3H)-one, 3-Hydroxy-4,4-dimethyloxolan-2-one, 3-Hydroxydihydro-4,4-dimethyl-2(3H)-furanone, DL-Pantoic acid γ-lactone, DL-Pantoyl lactone, NSC 135788, NSC 5926, dl-Pantoyl lactone, α-Hydroxy-β,β-dimethyl-γ-butyrolactone, α-Oxy-β,β-dimethyl-γ-butyrolactone |
| InChI Formula | InChI=1S/C6H10O3/c1-6(2)3-9-5(8)4(6)7/h4,7H,3H2,1-2H3 |
| SMILES | CC1(C)COC(=O)C1O |
| IUPAC Name | 3-hydroxy-4,4-dimethyloxolan-2-one |
| Molecular Weight (g/mol) | 130.14 |
| Formula Weight | 130.063 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction