missing translation for 'onlineSavingsMsg'
Learn More

D,L-Pantolactone, Mikromol™

Product Code. 30049114
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049114 25 mg 25mg
17969114 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30049114 Supplier Mikromol Supplier No. MM1352.040025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

D,L-Pantolactone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Pantolactone
CAS 79-50-5
Analyte or Component Names DL-Pantolactone
Shipping Condition Room Temperature
Molecular Formula C6 H10 O3
Certifications/Compliance ISO 9001
Synonym (±)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, D,L-Pantolactone, DL-Pantolactone, (+/-)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, Calcium Pantothenate Impurity C
InChI Formula InChI=1S/C6H10O3/c1-6(2)3-9-5(8)4(6)7/h4,7H,3H2,1-2H3
SMILES CC1(C)COC(=O)C1O
IUPAC Name 3-hydroxy-4,4-dimethyloxolan-2-one
Molecular Weight (g/mol) 130.14
Quantity 25 mg
Formula Weight 130.063 g/mol
Show More Show Less
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.