missing translation for 'onlineSavingsMsg'
Learn More

D,L-o-Tyrosine, TRC

Product Code. 30453288
Change view
Klicka för att se tillgängliga alternativ
Quantity:
100 mg
Förpackningsstorlek:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
30453288 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30453288 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT899980100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D,L-o-Tyrosine
CAS 2370-61-8
Molecular Formula C9 H11 N O3
Quantity 100 mg
Synonym Phenylalanine','2-hydroxy-, DL-Phenylalanine','2-hydroxy-, o-Tyrosine','DL- (8CI), 2-Hydroxyphenylalanine, (±)-o-Tyrosine, 2-Amino-3-(2-hydroxyphenyl)propanoic acid, 2-Azaniumyl-3-(2-hydroxyphenyl)propanoate, DL-2-Hydroxyphenylalanine, DL-o-Tyrosine, NSC 72345, Ortho-tyrosine, o-DL-Tyrosine, o-Tyrosine, DL-α-Amino-β-(o-hydroxyphenyl)propionic acid
InChI Formula InChI=1S/C9H11NO3/c10-7(9(12)13)5-6-3-1-2-4-8(6)11/h1-4,7,11H,5,10H2,(H,12,13)
SMILES NC(Cc1ccccc1O)C(=O)O
IUPAC Name 2-amino-3-(2-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol) 181.19
Formula Weight 181.0739
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.