missing translation for 'onlineSavingsMsg'
Learn More

D,L-O-Desmethyl Venlafaxine-d6, TRC

Product Code. 30348859
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348859 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30348859 Supplier Toronto Research Chemicals Supplier No. TRCD29650310MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material (±)-O-Desmethylvenlafaxine-D6
CAS 1062605-69-9
Purity Grade Notes HPLC
Molecular Formula C16 2H6 H19 N O2
Quantity 10 mg
Synonym O-Desmethylvenlafaxine-D6
InChI Formula InChI=1S/C16H25NO2/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16/h6-9,15,18-19H,3-5,10-12H2,1-2H3/i1D3,2D3
SMILES [2H]C([2H])([2H])N(CC(c1ccc(O)cc1)C2(O)CCCCC2)C([2H])([2H])[2H]
IUPAC Name 4-[2-[bis(trideuteriomethyl)amino]-1-(1-hydroxycyclohexyl)ethyl]phenol
Molecular Weight (g/mol) 269.41
Formula Weight 269.2262
Percent Purity >95
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.