missing translation for 'onlineSavingsMsg'
Learn More

D,L-N,N-Didesmethyl-O-desmethyl Venlafaxine, TRC

Product Code. 30374315
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30374315 25 mg -
30380585 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30374315 Supplier Toronto Research Chemicals Supplier No. TRCD44094525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149289-29-2
Molecular Formula C14 H21 N O2
Molecular Weight (g/mol) 235.32
Synonym Phenol, 4-[2-amino-1-(1-hydroxycyclohexyl)ethyl]-, 4-[2-Amino-1-(1-hydroxycyclohexyl)ethyl]phenol, 1-[2-Amino-1-(4-hydroxyphenyl)ethyl]cyclohexanol, O-Desmethyl-N,N-didesmethylvenlafaxine, Tridesmethylvenlafaxine
IUPAC Name 4-[2-amino-1-(1-hydroxycyclohexyl)ethyl]phenol
SMILES NCC(c1ccc(O)cc1)C2(O)CCCCC2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H21NO2/c15-10-13(11-4-6-12(16)7-5-11)14(17)8-2-1-3-9-14/h4-7,13,16-17H,1-3,8-10,15H2
Formula Weight 235.1572
Percent Purity >95%
Chemical Name or Material D,L-N,N-Didesmethyl-O-desmethyl Venlafaxine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.