missing translation for 'onlineSavingsMsg'
Learn More
Learn More
d,l-Metamfepramone.HCl, Lipomed
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
d,l-Metamfepramone.HCl
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethylcathinone (hydrochloride) |
| CAS | 10105-90-5 |
| Analyte or Component Names | N,N-Dimethylcathinone (hydrochloride) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H15 N O . Cl H |
| Synonym | Dimethylcathinone Hydrochloride, N,N-Dimethylcathinone Hydrochloride, Metamfepramone Hydrochloride, 1-Propanone, 2-(dimethylamino)-1-phenyl-, hydrochloride (1:1), 1-Propanone, 2-(dimethylamino)-1-phenyl-, hydrochloride (9 CI), Propiophenone, 2-(dimethylamino)-, hydrochloride (6 CI,7 CI,8 CI), Propiophenone, 2-(dimethylamino)-, hydrochloride (6 CI,7 CI,8 CI) |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CC(N(C)C)C(=O)c1ccccc1 |
| IUPAC Name | 2-(dimethylamino)-1-phenylpropan-1-one;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction