missing translation for 'onlineSavingsMsg'
Learn More

D,L-Homoglutamine, TRC

Product Code. 30390669
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390669 250 mg
30389997 100 mg
30393726 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390669 Supplier Toronto Research Chemicals Supplier No. TRCH593490250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34218-76-3
Molecular Formula C12 H24 N2 O6
Molecular Weight (g/mol) 292.329
IUPAC Name 6-amino-N-[(2R,5R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]hexanamide
SMILES NCCCCCC(=O)N[C@@H]1OC(CO)[C@H](O)C(O)C1O

Specifications

InChI Formula InChI=1S/C12H24N2O6/c13-5-3-1-2-4-8(16)14-12-11(19)10(18)9(17)7(6-15)20-12/h7,9-12,15,17-19H,1-6,13H2,(H,14,16)/t7?,9-,10?,11?,12+/m0/s1
Quantity 250 mg
Formula Weight 292.163
Shipping Condition Room Temperature
Chemical Name or Material D,L-Homoglutamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.