missing translation for 'onlineSavingsMsg'
Learn More

D,L-erythro-4'-Methyl-alpha-(1-isopropylaminoethyl) Benzyl Alcohol Hydrochloride, TRC

Product Code. 30179164
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179164 1 g 1g
30179009 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179164 Supplier Toronto Research Chemicals Supplier No. TRCM3145001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13549-69-4
Molecular Formula C13 H21 N O . Cl H
Molecular Weight (g/mol) 243.77
IUPAC Name (1S,2S)-1-(4-methylphenyl)-2-(propan-2-ylamino)propan-1-ol;hydrochloride
SMILES Cl.CC(C)N[C@@H](C)[C@@H](O)c1ccc(C)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H21NO.ClH/c1-9(2)14-11(4)13(15)12-7-5-10(3)6-8-12;/h5-9,11,13-15H,1-4H3;1H/t11-,13+;/m0./s1
Formula Weight 243.139
Chemical Name or Material D,L-erythro-4'-Methyl-alpha-(1-isopropylaminoethyl) Benzyl alcohol hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.