missing translation for 'onlineSavingsMsg'
Learn More

D(-)-isovaline, 97%

Product Code. 10645503 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10645503 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10645503 Supplier Thermo Scientific Acros Supplier No. 388032500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3059-97-0 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3059-97-0
Melting Point 276°C to 278°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C5H11NO2
Linear Formula CH3CH2CNH2(CH3)COOH
MDL Number MFCD00800497
Quantity 250 mg
Merck Index 15, 5280
Specific Rotation -9.0° to -12.0° (20°C, 589nm) (C=1, water)
Synonym d-isovaline, l-isovaline, s-2-amino-2-methylbutanoic acid, l-iso-valine, unii-ebv7h5w26h, ebv7h5w26h, isovaline, l, d--isovaline, 2s-2-amino-2-methylbutanoic acid, s-2-amino-2-methylbutyric acid
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol,slightly soluble in ether
InChI Key GCHPUFAZSONQIV-RXMQYKEDSA-N
SMILES CC[C@@](C)([NH3+])C([O-])=O
IUPAC Name (2S)-2-amino-2-methylbutanoic acid
Molecular Weight (g/mol) 117.15
PubChem CID 6971276
Formula Weight 117.15
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material D(-)-Isovaline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.