missing translation for 'onlineSavingsMsg'
Learn More

D-Isoleucine Methyl Ester Hydrochloride, TRC

Product Code. 30400942
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400942 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400942 Supplier Toronto Research Chemicals Supplier No. TRCI785060100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl (2R,3R)-2-Amino-3-methylpentanoate Hydrochloride
CAS 167223-42-9
Molecular Formula C7 H15 N O2 . Cl H
Quantity 100 mg
Synonym D-Isoleucine Methyl Ester Hydrochloride, D-Isoleucine, methyl ester, hydrochloride (1:1), D-Isoleucine, methyl ester, hydrochloride (9CI), Methyl D-isoleucinate hydrochloride, D-Isoleucine methyl ester hydrochloride, Methyl (2R,3R)-2-amino-3-methylpentanoate hydrochloride, (2R,3R)-2-Amino-3-meth
InChI Formula InChI=1S/C7H15NO2.ClH/c1-4-5(2)6(8)7(9)10-3;/h5-6H,4,8H2,1-3H3;1H/t5-,6-;/m1./s1
SMILES Cl.CC[C@@H](C)[C@@H](N)C(=O)OC
IUPAC Name methyl (2R,3R)-2-amino-3-methylpentanoate;hydrochloride
Molecular Weight (g/mol) 181.66
Formula Weight 181.087
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.